trans-Bis(triphenylphosphin)palladium(II)chlorid ≥99%

Lieferant: abcr

Synonyme: trans-Dichlorobis(triphenylphosphine)palladium(II)

AB116703.0001
ABCRAB116703.0001EA 69.9 EUR
ABCRAB116703.0001
trans-Bis(triphenylphosphin)palladium(II)chlorid ≥99%
trans-Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
MDL: MFCD00009593
CAS-Nummer: 13965-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR