(4-Fluorbenzyl)triphenylphosphoniumchlorid ≥98%

Lieferant: ABCR
Achtung

AB116807.0100
ABCRAB116807.0100EA 0 EUR
ABCRAB116807.0100
(4-Fluorbenzyl)triphenylphosphoniumchlorid ≥98%
(4-Fluorbenzyl)triphenylphosphoniumchlorid

Purity: 98+% CAS No.: 3462-95-1 Molecular Formula: C25H21ClFP Formula Weight: 406.87 Melting Point: 310-312° Beilstein Reference: 4075331

Formel: C₂₅H₂₁ClFP
Molecular Weight: 406,87 g/mol
Schmelzpunkt: 310…312 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031637
CAS-Nummer: 3462-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR