2-Hydroxynicotinsäure ≥99%
Lieferant: abcr
|
Achtung
|
AB117297.0100
ABCRAB117297.0100EA
0
EUR
ABCRAB117297.0100
2-Hydroxynicotinsäure ≥99%
2-Hydroxynicotinsäure
Purity: 99%
CAS No.: 609-71-2
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Melting Point: 256-261°
Beilstein Reference: 119028
Formel:
C₆H₅NO₃ Schmelzpunkt: 256…261 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010100 CAS-Nummer: 609-71-2 EINECS: 210-198-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...