3-(Trimethoxysilyl)propylmethacrylat ≥97%
Lieferant: Thermo Fisher Scientific
A17714.22
A17714.36
AB117674.0100
A17714.22EA
45.4
EUR
A17714.22
A17714.36
ABCRAB117674.0100
3-(Trimethoxysilyl)propylmethacrylat ≥97%
3-(Trimethoxysilyl)propylmethacrylat
Purity: 97%
CAS No.: 2530-85-0
Molecular Formula: C10H20O5Si
Formula Weight: 248.35
Flash Point: 92°(197°F)
Refractive Index: 1.431
Boiling Point: 190°
Beilstein Reference: 1952435
MDL No.: MFCD00008593
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...