2-(Trifluormethoxy)benzoesäure ≥98%

Lieferant: ABCR
Achtung

AB119307.0001
ABCRAB119307.0001EA 0 EUR
ABCRAB119307.0001
2-(Trifluormethoxy)benzoesäure ≥98%
2-(Trifluormethoxy)benzoesäure

Purity: 98% CAS No.: 1979-29-9 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Melting Point: 78-80° Beilstein Reference: 2645481

Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052325
CAS-Nummer: 1979-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR