Mangan(III)acetat Dihydrat ≥98%

Lieferant: ABCR
Achtung

Synonyme: Essigsäure Mangan(III)salz Dihydrat, Mangantriacetat Dihydrat

AB120967.0025
ABCRAB120967.0025EA 0 EUR
ABCRAB120967.0025
Mangan(III)acetat Dihydrat ≥98%
Mangan(III)acetat Dihydrat
Formel: (H₃CCOO)₂Mn·2H₂O
Molecular Weight: 209,06 g/mol
MDL: MFCD00150022
CAS-Nummer: 19513-05-4
EINECS: 213-602-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR