Molybdän(VI)oxid ≥99,998% (Mo-Basis), PURATREM

Lieferant: ABCR
Achtung

Synonyme: Molybdenum trioxide

AB121057.0050
ABCRAB121057.0050EA 0 EUR
ABCRAB121057.0050
Molybdän(VI)oxid ≥99,998% (Mo-Basis), PURATREM
Molybdenum(VI) oxide
Formel: MoO₃
Molecular Weight: 143,94 g/mol
Siedepunkt: (Subl.)
Schmelzpunkt: 795 °C
Dichte: 4,69
MDL: MFCD00003469
CAS-Nummer: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 13,06261

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR