Palladium(II)bromid ≥99%

Lieferant: abcr
Achtung

AB121379.0005
ABCRAB121379.0005EA 0 EUR
ABCRAB121379.0005
Palladium(II)bromid ≥99%
Palladium(II)bromid
Formel: Br₂Pd
Dichte: 5,173
MDL: MFCD00011170
CAS-Nummer: 13444-94-5
EINECS: 236-588-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR