(1-Hexadecyl)tributylphosphoniumbromid ≥98%

Lieferant: ABCR
Achtung

Synonyme: Hexadecyltri-n-butylphosphonium bromide, Tributylhexadecylphosphonium bromide, Tributyl(hexadecyl)phosphonium bromide, (n-Hexadecyl)tri-n-butylphosphonium bromide

AB121580.0010
ABCRAB121580.0010EA 0 EUR
ABCRAB121580.0010
(1-Hexadecyl)tributylphosphoniumbromid ≥98%
(1-Hexadecyl)tributylphosphoniumbromid
Formel: C₂₈H₆₀BrP
Molecular Weight: 507,65 g/mol
Schmelzpunkt: 54…56 °C
MDL: MFCD00011775
CAS-Nummer: 14937-45-2
EINECS: 239-014-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR