Tetraphenylphosphoniumbromid ≥99%

Lieferant: abcr
Achtung

AB121614.0025
ABCRAB121614.0025EA 0 EUR
ABCRAB121614.0025
Tetraphenylphosphoniumbromid ≥99%
Tetraphenylphosphonium bromide
Formel: C₂₄H₂₀BrP
Schmelzpunkt: 294…296 °C
MDL: MFCD00011915
CAS-Nummer: 2751-90-8
EINECS: 220-393-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR