Ytterbiumtriacetat Hydrat ≥99,9% (Yb-Basis)

Lieferant: ABCR
Achtung

Synonyme: Ytterbium(III) acetate hydrate

AB122261.0010
ABCRAB122261.0010EA 0 EUR
ABCRAB122261.0010
Ytterbiumtriacetat Hydrat ≥99,9% (Yb-Basis)
Ytterbiumtriacetat Hydrat
Formel: C₆H₉O₆Yb·H₂O
Molecular Weight: 368,19 g/mol
Dichte: 2,09
MDL: MFCD00150127
CAS-Nummer: 15280-58-7
EINECS: 244-137-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR