2-(1-Naphthoxy)ethanol

Lieferant: ABCR

AB123307.0005
ABCRAB123307.0005EA 0 EUR
ABCRAB123307.0005
2-(1-Naphthoxy)ethanol
2-(1-Naphthoxy)ethanol
Formel: C₁₂H₁₂O₂
CAS-Nummer: 711-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR