4-Acetamido-3-chlorbenzolsulfonylchlorid ≥97%
Lieferant: ABCR
AB124858.0005
ABCRAB124858.0005EA
0
EUR
ABCRAB124858.0005
4-Acetamido-3-chlorbenzolsulfonylchlorid ≥97%
4-Acetamido-3-chlorbenzolsulfonylchlorid
Purity: 97%
CAS No.: 16761-18-5
Molecular Formula: C8H7Cl2NO3S
Formula Weight: 268.12
Storage Temperature: 2°C to 8°C
Melting Point: ca 140° dec.
UN No.: 3261
MDL No.: MFCD00052363
Formel:
C₈H₇Cl₂NO₃S |
MDL:
MFCD00052363 CAS-Nummer: 16761-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...