7-Nitroindol ≥97%

Lieferant: ABCR
Achtung

AB124915.0001
ABCRAB124915.0001EA 0 EUR
ABCRAB124915.0001
7-Nitroindol ≥97%
7-Nitroindol

Purity: 97% CAS No.: 6960-42-5 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Odour: Odourless Melting Point: 94-96° Beilstein Reference: 130874 MDL No.: MFCD00005683

Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005683
CAS-Nummer: 6960-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR