1-Amino-2,4-dibrom-9,10-anthrachinon ≥97%

Lieferant: ABCR
Achtung

Synonyme: 1-Amino-2,4-dibromoanthraquinone

AB124984.0250
ABCRAB124984.0250EA 0 EUR
ABCRAB124984.0250
1-Amino-2,4-dibrom-9,10-anthrachinon ≥97%
1-Amino-2,4-dibrom-9,10-anthrachinon

Purity: 97% CAS No.: 81-49-2 Molecular Formula: C14H7Br2NO2 Formula Weight: 381.03 Odour: Odourless Melting Point: 225-229° MDL No.: MFCD00019155

Formel: C₁₄H₇Br₂NO₂
Molecular Weight: 381,03 g/mol
Schmelzpunkt: 225…229 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019155
CAS-Nummer: 81-49-2
EINECS: 201-354-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR