Methyl-3,4-dimethylbenzoat ≥98%

Lieferant: ABCR

Synonyme: Methyl 3,4-dimethylbenzoate

AB125607.0250
ABCRAB125607.0250EA 0 EUR
ABCRAB125607.0250
Methyl-3,4-dimethylbenzoat ≥98%
Methyl-3,4-dimethylbenzoat

Purity: 98% CAS No.: 38404-42-1 Molecular Formula: C10H12O2 Formula Weight: 164.21 Melting Point: 35-37° MDL No.: MFCD00219861

Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2011 g/mol
Schmelzpunkt: 35…37 °C
MDL: MFCD00219861
CAS-Nummer: 38404-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR