(2E)-2-Dodecen-1-ol

Lieferant: ABCR

Synonyme: trans-2-Dodecenol

AB126394.0025
ABCRAB126394.0025EA 0 EUR
ABCRAB126394.0025
(2E)-2-Dodecen-1-ol
trans-2-Dodecen-1-ol
Formel: C₁₂H₂₄O
Molecular Weight: 184,3184 g/mol
MDL: MFCD00014047
CAS-Nummer: 69064-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR