4,5-Dibrom-2-thenoesäure ≥98%

Lieferant: ABCR
Achtung

Synonyme: 4,5-Dibromthiophen-2-carbonsäure, 4,5-Dibrom-2-thiophencarbonsäure

AB126611.0100
ABCRAB126611.0100EA 0 EUR
ABCRAB126611.0100
4,5-Dibrom-2-thenoesäure ≥98%
4,5-Dibrom-2-thenoesäure

Purity: 98% CAS No.: 6324-10-3 Molecular Formula: C5H2Br2O2S Formula Weight: 285.94 Odour: Odourless Melting Point: 228-232° Beilstein Reference: 135885 MDL No.: MFCD00039682

Formel: C₅H₂Br₂O₂S
Molecular Weight: 285,94 g/mol
Schmelzpunkt: 228…232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039682
CAS-Nummer: 6324-10-3
EINECS: 228-683-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR