4-Fluor-3-methoxybenzonitril

Lieferant: ABCR

Synonyme: 4-Fluor-3-methoxybenzoesäurenitril

AB129485.0100
ABCRAB129485.0100EA 0 EUR
ABCRAB129485.0100
4-Fluor-3-methoxybenzonitril
4-Fluor-3-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD01569371
CAS-Nummer: 243128-37-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR