4-Piperidinol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Piperidin-4-ol, 4-Hydroxypiperidin

B22723.06 B22723.14 B22723.22 AB131656.0025 B22723.02
B22723.06EA 34.2 EUR
B22723.06 B22723.14 B22723.22 ABCRAB131656.0025 B22723.02
4-Piperidinol ≥97%
4-Piperidinol

Purity: 97% CAS No.: 5382-16-1 Molecular Formula: C5H11NO Formula Weight: 101.15 Flash Point: 107°(224°F) Odour: Amine-like pH: 11.5-11.7 Melting Point: 86-90° Boiling Point: 108-114°/10mm Beilstein Reference: 102738 UN No.: 3263 MDL No.: MFCD00005999

Formel: C₅H₁₁NO
Molecular Weight: 101.15 g/mol
Siedepunkt: 108…114 °C (10 mmHg)
Schmelzpunkt: 86…90 °C
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005999
CAS-Nummer: 5382-16-1
EINECS: 226-373-1
UN: 3263
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR