5-Brom-2,2-difluor-1,3-benzodioxol ≥97%
Lieferant: ABCR
|
Achtung
|
Synonyme:
5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole
AB131838.0005
ABCRAB131838.0005EA
0
EUR
ABCRAB131838.0005
5-Brom-2,2-difluor-1,3-benzodioxol ≥97%
5-Brom-2,2-difluor-1,3-benzodioxol
Formel:
C₇H₃BrF₂O₂ Molecular Weight: 237 g/mol Siedepunkt: 78…79 °C (20 mmHg) Dichte: 1,740 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236212 CAS-Nummer: 33070-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...