β-Chlor-4'-fluorpropiophenon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone

B21891.06 B21891.14 B21891.22 AB132433.0100
B21891.06EA 25.9 EUR
B21891.06 B21891.14 B21891.22 ABCRAB132433.0100
β-Chlor-4'-fluorpropiophenon ≥97%
β-Chlor-4'-fluorpropiophenon

Purity: 97% CAS No.: 347-93-3 Molecular Formula: C9H8ClFO Formula Weight: 186.61 Melting Point: 46-49° Boiling Point: 100-103°/3mm MDL No.: MFCD00000991

Formel: C₉H₈ClFO
Molecular Weight: 186,61 g/mol
Siedepunkt: 100…103 °C (3 mmHg)
Schmelzpunkt: 46…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000991
CAS-Nummer: 347-93-3
EINECS: 206-475-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR