1-(2-Methylphenyl)-2-thioharnstoff ≥97%

Lieferant: ABCR
Gefahr

Synonyme: 2-Methylphenylthioharnstoff, 1-(2-Tolyl)thioharnstoff, o-Tolylthioharnstoff, N-(2-Methylphenyl)thioharnstoff

AB132998.0050
ABCRAB132998.0050EA 0 EUR
ABCRAB132998.0050
1-(2-Methylphenyl)-2-thioharnstoff ≥97%
1-(2-Methylphenyl)-2-thioharnstoff

Purity: 97+% CAS No.: 614-78-8 Molecular Formula: C8H10N2S Formula Weight: 166.25 Odour: Odourless Melting Point: 155-158° Beilstein Reference: 2086802 UN No.: 2811 MDL No.: MFCD00041162

Formel: (C₆H₄(CH₃))NC(S)NH₂
Molecular Weight: 166,25 g/mol
Siedepunkt: 295 °C (1013 hPa)
Schmelzpunkt: 155…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041162
CAS-Nummer: 614-78-8
EINECS: 210-395-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR