1,1,3,3-Tetramethyldisiloxan 97%

Lieferant: ABCR
Gefahr

AB133126.0100
ABCRAB133126.0100EA 0 EUR
ABCRAB133126.0100
1,1,3,3-Tetramethyldisiloxan 97%
1,1,3,3-Tetramethyldisiloxan
Formel: C₄H₁₄OSi₂
Molecular Weight: 134,33 g/mol
MDL: MFCD00008256
CAS-Nummer: 3277-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR