α-Brom-2-iodtoluol ≥96%

Lieferant: abcr
Gefahr

Synonyme: α-Brom-o-iodtoluol, 2-Iodbenzylbromid

AB133194.0005
ABCRAB133194.0005EA 0 EUR
ABCRAB133194.0005
α-Brom-2-iodtoluol ≥96%
α-Brom-2-iodtoluol

Purity: 96% CAS No.: 40400-13-3 Molecular Formula: C7H6BrI Formula Weight: 296.94 Melting Point: 53-58° Beilstein Reference: 2079736 UN No.: 3261 MDL No.: MFCD00236046

Formel: C₇H₆BrI
Molecular Weight: 296.93 g/mol
Siedepunkt: 279
Schmelzpunkt: 55,5
MDL: MFCD00236046
CAS-Nummer: 40400-13-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR