2-(Difluormethoxy)benzaldehyd ≥97%

Lieferant: ABCR
Achtung

AB133792.0005
ABCRAB133792.0005EA 0 EUR
ABCRAB133792.0005
2-(Difluormethoxy)benzaldehyd ≥97%
2-(Difluormethoxy)benzaldehyd

Purity: 97% CAS No.: 71653-64-0 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Flash Point: 106°(223°F) Refractive Index: 1.4970 Boiling Point: 88-90°/9mm Beilstein Reference: 389068 MDL No.: MFCD00042251

Formel: C₈H₆F₂O₂
Molecular Weight: 172,13 g/mol
Siedepunkt: 88…90 °C (9 mmHg)
Dichte: 1,284
Flammpunkt: 106 °C (223 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042251
CAS-Nummer: 71653-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR