2-(Difluormethoxy)benzoesäure ≥97%

Lieferant: abcr
Achtung

AB133793.0001
ABCRAB133793.0001EA 0 EUR
ABCRAB133793.0001
2-(Difluormethoxy)benzoesäure ≥97%
2-(Difluormethoxy)benzoesäure

Purity: 97% CAS No.: 97914-59-5 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 98-100° MDL No.: MFCD00236223

Formel: C₈H₆F₂O₃
Molecular Weight: 188.13 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236223
CAS-Nummer: 97914-59-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR