4-(Methylsulfonyl)anilin Hydrochlorid ≥99%
Lieferant: abcr
|
Gefahr
|
AB134063.0025
ABCRAB134063.0025EA
0
EUR
ABCRAB134063.0025
4-(Methylsulfonyl)anilin Hydrochlorid ≥99%
4-(Methylsulfonyl)anilin Hydrochlorid
Formel:
C₇H₁₀ClNO₂S Schmelzpunkt: 237…239 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216490 CAS-Nummer: 177662-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...