(±)-1,2-Hexandiol
Lieferant: abcr
Synonyme:
(±)-1,2-Dihydroxyhexan
AB135150.0025
ABCRAB135150.0025EA
0
EUR
ABCRAB135150.0025
(±)-1,2-Hexandiol
(±)-1,2-Hexandiol
Formel:
C₆H₁₄O₂ Molecular Weight: 118.18 g/mol Siedepunkt: 223 Schmelzpunkt: 45 Dichte: 0.9507 g/cm³ (20 °C) |
MDL:
MFCD00010737 CAS-Nummer: 6920-22-5 EINECS: 230-029-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...