16-Hentriacontanon

Lieferant: abcr

AB135197.0025
ABCRAB135197.0025EA 0 EUR
ABCRAB135197.0025
16-Hentriacontanon
16-Hentriacontanon
Formel: C₃₁H₆₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00059222
CAS-Nummer: 502-73-8
EINECS: 207-952-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR