Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid)
Lieferant: ABCR
Synonyme:
2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride
AB135314.0025
ABCRAB135314.0025EA
0
EUR
ABCRAB135314.0025
Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid)
Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid)
Formel:
C₅H₁₄ClN₃O Molecular Weight: 167,64 g/mol |
MDL:
MFCD00012009 CAS-Nummer: 123-46-6 EINECS: 204-629-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...