cis-4-Aminocyclohexancarbonsäure

Lieferant: ABCR

AB135321.0005
ABCRAB135321.0005EA 0 EUR
ABCRAB135321.0005
cis-4-Aminocyclohexancarbonsäure
cis-4-Aminocyclohexanecarboxylic acid
Formel: C₇H₁₃NO₂
Molecular Weight: 143,19 g/mol
MDL: MFCD00191730
CAS-Nummer: 3685-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR