3'-Amino-4'-methoxyacetanilid ≥98,0% (durch HPLC, titrimetrische Analyse)
Lieferant: abcr
Synonyme:
5-Acetamido-o-anisidine, 2-Amino-4-acetamino anisole, N1-Acetyl-4-methoxy-1,3-phenylenediamine, N-(3-Amino-4-methoxyphenyl)acetamide
AB136303.0500
ABCRAB136303.0500EA
0
EUR
ABCRAB136303.0500
3'-Amino-4'-methoxyacetanilid ≥98,0% (durch HPLC, titrimetrische Analyse)
3'-Amino-4'-methoxyacetanilid
Formel:
C₉H₁₂N₂O₂ |
MDL:
MFCD00008676 CAS-Nummer: 6375-47-9 EINECS: 228-938-8 |
|
Spezifikation Testergebnisse
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...