1-Amino-2-methylnaphthalin

Lieferant: ABCR

Synonyme: 2-Methyl-1-naphthylamin

AB136336.0005
ABCRAB136336.0005EA 0 EUR
ABCRAB136336.0005
1-Amino-2-methylnaphthalin
1-Amino-2-methylnaphthalene
Formel: C₁₁H₁₁N
Molecular Weight: 157,22 g/mol
MDL: MFCD00059135
CAS-Nummer: 2246-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR