6-Amino-1-naphthalinsulfonsäure ≥97,0% (durch HPLC, titrimetrische Analyse)
Lieferant: ABCR
|
Achtung
|
Synonyme:
6-Amino-1-naphthalenesulphonic acid
AB136367.0500
ABCRAB136367.0500EA
0
EUR
ABCRAB136367.0500
6-Amino-1-naphthalinsulfonsäure ≥97,0% (durch HPLC, titrimetrische Analyse)
6-Amino-1-naphthalenesulphonic acid
Formel:
C₁₀H₉NO₃S Molecular Weight: 223,2500 g/mol |
MDL:
MFCD00035724 CAS-Nummer: 81-05-0 EINECS: 201-318-4 Merck Index: 14,06407 |
Spezifikation Testergebnisse
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...