3-Hydroxyanthranilsäure

Lieferant: ABCR

Synonyme: 3-Hydroxyanthranilic acid

AB136832
ABCRAB136832EA 0 EUR
ABCRAB136832
3-Hydroxyanthranilsäure
3-Hydroxyanthranilsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Siedepunkt: 357 °C (1013 hPa)
Schmelzpunkt: 164 °C
MDL: MFCD00007700
CAS-Nummer: 548-93-6
EINECS: 208-962-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR