1-Hydroxy-9,10-anthrachinon ≥98,0% (durch HPLC)

Lieferant: ABCR
Achtung

Synonyme: 1-Hydroxyanthraquinone

AB136834.0025
ABCRAB136834.0025EA 266 EUR
ABCRAB136834.0025
1-Hydroxy-9,10-anthrachinon ≥98,0% (durch HPLC)
1-Hydroxy-9,10-anthrachinon
Formel: C₁₄H₈O₃
Molecular Weight: 224,2115 g/mol
MDL: MFCD00058946
CAS-Nummer: 129-43-1
EINECS: 204-946-7

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR