6-Hydroxy-1-naphthoesäure

Lieferant: abcr

Synonyme: 6-Hydroxy-1-naphthalenecarboxylic acid

AB136999.0005
ABCRAB136999.0005EA 0 EUR
ABCRAB136999.0005
6-Hydroxy-1-naphthoesäure
6-Hydroxy-1-naphthoesäure
Formel: C₁₁H₈O₃
MDL: MFCD00209983
CAS-Nummer: 2437-17-4
EINECS: 407-390-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR