2-Mercaptopyridin-N-oxid Zinksalz

Lieferant: ABCR

Synonyme: Bis(2-pyridylthio) Zinc 1,1'-Dioxide, Zinc Pyrithione, 2-Pyridinethiol 1-Oxide Zinc salt, 1-Hydroxypyridine-2-thione Zinc salt

AB137921
ABCRAB137921EA 0 EUR
ABCRAB137921
2-Mercaptopyridin-N-oxid Zinksalz
2-Mercaptopyridin-N-oxid Zinksalz
Formel: C₁₀H₈N₂O₂S₂Zn
Molecular Weight: 317,72 g/mol
MDL: MFCD00067336
CAS-Nummer: 13463-41-7
EINECS: 236-671-3
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR