(4-Methoxyphenyl)essigsäureethylester

Lieferant: ABCR

Synonyme: Ethyl-(4-methoxyphenyl)acetat

AB138338.0025
ABCRAB138338.0025EA 0 EUR
ABCRAB138338.0025
(4-Methoxyphenyl)essigsäureethylester
(4-Methoxyphenyl)essigsäureethylester
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Siedepunkt: 138 °C (1013 hPa)
Dichte: 1,097 g/cm³ (24 °C)
MDL: MFCD00040760
CAS-Nummer: 14062-18-1
EINECS: 237-903-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR