Boc-Pro-OH

Lieferant: abcr

Synonyme: N-(tert-Butoxycarbonyl)-L-proline, N-Boc-L-proline, N-α-t-Boc-L-Proline, BOC-L-Proline, (S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid

AB138550.0005
ABCRAB138550.0005EA 0 EUR
ABCRAB138550.0005
Boc-Pro-OH
Boc-Pro-OH
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215.25 g/mol
MDL: MFCD00037324
CAS-Nummer: 15761-39-4
EINECS: 239-848-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR