2-(2-(2-Chlorethoxy)ethoxy)ethanol

Lieferant: ABCR
Achtung

AB139597.0025
ABCRAB139597.0025EA 0 EUR
ABCRAB139597.0025
2-(2-(2-Chlorethoxy)ethoxy)ethanol
2-(2-(2-Chloroethoxy)ethoxy)ethanol
Formel: C₆H₁₃ClO₃
Molecular Weight: 168,62 g/mol
MDL: MFCD00002874
CAS-Nummer: 5197-62-6
EINECS: 225-984-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR