9-Phenylacridin

Lieferant: abcr

AB140354.0010
ABCRAB140354.0010EA 0 EUR
ABCRAB140354.0010
9-Phenylacridin
9-Phenylacridine
Formel: C₁₉H₁₃N
Molecular Weight: 255.32 g/mol
MDL: MFCD00092562
CAS-Nummer: 602-56-2
EINECS: 210-020-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR