β-Cyclodextrin

Lieferant: ABCR

Synonyme: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

AB140413.0025
ABCRAB140413.0025EA 0 EUR
ABCRAB140413.0025
β-Cyclodextrin
β-Cyclodextrin
Formel: C₄₂H₇₀O₃₅
Molecular Weight: 1134,98 g/mol
MDL: MFCD00078139
CAS-Nummer: 7585-39-9
EINECS: 231-493-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR