Chrom(III)picolinat ≥96,0% (durch titrimetrische Analyse)
Lieferant: abcr
|
Achtung
|
Synonyme:
Picolinic acid chromium salt, Chromium picolinate
AB141096.0025
ABCRAB141096.0025EA
0
EUR
ABCRAB141096.0025
Chrom(III)picolinat ≥96,0% (durch titrimetrische Analyse)
Chrom(III)picolinat
Formel:
C₁₈H₁₂CrN₃O₆ |
MDL:
MFCD00068715 CAS-Nummer: 14639-25-9 Merck Index: 14,02236 |
Spezifikation Testergebnisse
Appearance | Purple to Dark purple to Dark red powder to crystal |
Purity (Iodometric Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...