Pyridine-2-carbohydrazide ≥98,0% (durch titrimetrische Analyse)

Lieferant: abcr
Achtung

AB141104.0025
ABCRAB141104.0025EA 0 EUR
ABCRAB141104.0025
Pyridine-2-carbohydrazide ≥98,0% (durch titrimetrische Analyse)
Pyridine-2-carbohydrazide
Formel: C₆H₇N₃O
MDL: MFCD00059782
CAS-Nummer: 1452-63-7

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 98.0 %
Melting point 99.0 to 102.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR