N,N'-Di-sec-butyl-p-phenylendiamin

Lieferant: ABCR

Synonyme: N,N'-Di-sec-butyl-1,4-phenylenediamine, N1,N4-Di-sec-butylbenzene-1,4-diamine

AB141113.0025
ABCRAB141113.0025EA 0 EUR
ABCRAB141113.0025
N,N'-Di-sec-butyl-p-phenylendiamin
N,N'-Di-sec-butyl-p-phenylendiamin
Formel: C₁₄H₂₄N₂
Molecular Weight: 220,3500 g/mol
MDL: MFCD00043658
CAS-Nummer: 101-96-2
EINECS: 202-992-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR