2,6-Dimethylhydrochinon

Lieferant: ABCR

Synonyme: 2,5-Dihydroxy-m-xylene

AB142400.0005
ABCRAB142400.0005EA 0 EUR
ABCRAB142400.0005
2,6-Dimethylhydrochinon
2,6-Dimethylhydrochinon
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
MDL: MFCD00016466
CAS-Nummer: 654-42-2
EINECS: 211-505-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR