2,5-Dimethyl-p-phenylendiamin

Lieferant: abcr

Synonyme: 2,5-Dimethylbenzol-1,4-diamin, 2,5-Dimethyl-1,4-phenylendiamin, 1,4-Diamino-2,5-dimethylbenzol

AB142491.0025
ABCRAB142491.0025EA 0 EUR
ABCRAB142491.0025
2,5-Dimethyl-p-phenylendiamin
2,5-Dimethyl-p-phenylendiamin
Formel: C₈H₁₂N₂
Molecular Weight: 136.2 g/mol
MDL: MFCD00052675
CAS-Nummer: 6393-01-7
EINECS: 229-003-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR