Toluendiisocyanat ≥99% (2,4-80%, 2,6-20%)

Lieferant: abcr
Gefahr

Synonyme: Toluylene diisocyanate

AB142608.0500
ABCRAB142608.0500EA 146 EUR
ABCRAB142608.0500
Toluendiisocyanat ≥99% (2,4-80%, 2,6-20%)
Toluendiisocyanat
Formel: C₉H₆N₂O₂
Molecular Weight: 174.16
Lagertemperatur: Raumtemperatur
MDL: MFCD01778251
CAS-Nummer: 26471-62-5
EINECS: 247-722-4
UN: 2078
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR